missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3CG Monoclonal Antibody (OTI4B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505255
This item is not returnable.
View return policy
Description
This gene encodes a protein that belongs to the pi3/pi4-kinase family of proteins. The gene product is an enzyme that phosphorylates phosphoinositides on the 3-hydroxyl group of the inositol ring. It is an important modulator of extracellular signals, including those elicited by E-cadherin-mediated cell-cell adhesion, which plays an important role in maintece of the structural and functional integrity of epithelia. In addition to its role in promoting assembly of adherens junctions, the protein is thought to play a pivotal role in the regulation of cytotoxicity in NK cells. The gene is located in a commonly deleted segment of chromosome 7 previously identified in myeloid leukemias.Specifications
| PIK3CG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P48736 | |
| PIK3CG | |
| Full length human recombit protein of human PIK3CG produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4B1 | |
| Unconjugated | |
| PIK3CG | |
| PI3CG, PI3K, PI3Kgamma, PIK3, p110gamma, p120-PI3K | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5294 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction