missing translation for 'onlineSavingsMsg'
Learn More

PIK3IP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. p-7208242 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16836584 20 μL 20µL
16826584 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16836584 Supplier Proteintech Supplier No. 168261AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The exact function of PIK3IP1 remains unknown.

Specifications

Antigen PIK3IP1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PIK3IP1
Gene Accession No. Q56A20, Q7TMJ8, Q96FE7
Gene Alias HGFL, hHGFL(S), PIK3IP1
Gene Symbols Pik3ip1
Host Species Rabbit
Immunogen PIK3IP1 Fusion Protein Ag10427
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113791, 216505, 305472
Target Species Human, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.