missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3R1, Rabbit anti-Human, APC, Clone: 1A11, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB19010.10Reactions
This item is not returnable.
View return policy
Specifications
| PIK3R1 | |
| Monoclonal | |
| APC | |
| In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide). | |
| GRB1/p85/p85-ALPHA | |
| Rabbit | |
| 10 Reactions | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| 1A11 | |
| Flow Cytometry (5 uL/106 cells or 0.05 ug/mL) The optimal working dilution should be determined by the end user. | |
| phosphoinositide-3-kinase, regulatory subunit 1 (alpha) | |
| PIK3R1 | |
| A synthetic phosphopeptide corresponding to residues surrounding Y458 of human PIK3R1. | |
| RUO | |
| 5295/8503 | |
| Store at 2-8°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction