missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIK3R2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586504
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
Binds to activated (phosphorylated) protein-tyrosine kinases, through its SH2 domain, and acts as an adapter, mediating the association of the p110 catalytic unit to the plasma membrane.
Specifications
| PIK3R2 | |
| Polyclonal | |
| Unconjugated | |
| PIK3R2 | |
| MPPH; MPPH1; p85; P85B; p85beta; p85-BETA; phosphatidylinositol 3-kinase; phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta; phosphatidylinositol 3-kinase regulartory subunit polypeptide 2; phosphatidylinositol 3-kinase regulatory subunit beta; phosphatidylinositol 3-kinase, regulartory subunit, polypeptide 2; phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 2 (p85 beta); phosphoinositide-3-kinase regulatory subunit 2; phosphoinositide-3-kinase regulatory subunit beta; phosphoinositide-3-kinase, regulatory subunit 2 (beta); phosphoinositide-3-kinase, regulatory subunit 2 (p85 beta); phosphoinositide-3-kinase, regulatory subunit, polypeptide 2 (p85 beta); PI3K; PI3K regulatory subunit beta; PI3-kinase p85 subunit beta; PI3-kinase regulatory subunit beta; PI3-kinase subunit p85-beta; PIK3R2; ptdIns-3-kinase p85-beta; ptdIns-3-kinase regulatory subunit beta; ptdIns-3-kinase regulatory subunit p85-beta | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18709, 29741, 5296 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00459, O08908, Q63788 | |
| PIK3R2 | |
| Synthetic peptide, corresponding to amino acids 471-520 of Human PI3K p85beta. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction