missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3R5 Monoclonal Antibody (OTI2E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505828S
This item is not returnable.
View return policy
Description
PIK3R5 Monoclonal antibody specifically detects PIK3R5 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| PIK3R5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WYR1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23533 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2E4 | |
| Unconjugated | |
| PIK3R5 | |
| AV230647; F730038I15Rik; FOAP-2; p101; p101-PI3K; phosphatidylinositol 4,5-bisphosphate 3-kinase regulatory subunit; phosphatidylinositol-4,5-bisphosphate 3-kinase regulatory subunit; phosphoinositide 3-kinase regulatory subunit 5; phosphoinositide-3-kinase regulatory subunit 5; phosphoinositide-3-kinase, regulatory subunit 5; phosphoinositide-3-kinase, regulatory subunit 5, p101; PI3-kinase p101 subunit; PI3-kinase regulatory subunit 5; Pik3r5; Protein FOAP-2; ptdIns-3-kinase p101; PtdIns-3-kinase regulatory subunit; RGD1563261 | |
| Full length human recombinant protein of human PIK3R5 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction