missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIKFYVE Mouse anti-Human, Clone: OTI1D2, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810374
This item is not returnable.
View return policy
Description
PIKFYVE belongs to a large family of lipid kinases that alter the phosphorylation status of intracellular phosphatidylinositol. The content of phosphatidylinositol 3,5-bisphosphateP2) in endosomal membranes changes dynamically with fission and fusion events that generate or absorb intracellular transport vesicles. PIKFYVE is the PtdIns(3,5)P2-producing component of a trimolecular complex that tightly regulates the level of PtdIns(3,5)P2. Other components of this complex are the PIKFYVE activator VAC14 and the PtdIns(3,5)P2 phosphatase FIG4. In addition to its phosphoinositide 5-kinase activity, PIKFYVE also has protein kinase activity. For a review, see Shisheva.Specifications
| PIKFYVE | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y2I7 | |
| PIKFYVE | |
| Full length human recombit protein of human PIKFYVE produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D2 | |
| Unconjugated | |
| PIKFYVE | |
| CFD, FAB1, HEL37, PIP5K, PIP5K3, ZFYVE29 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 200576 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction