missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIKFyve/PIP5K3 Antibody (1D11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00200576-M02
This item is not returnable.
View return policy
Description
PIKFyve/PIP5K3 Monoclonal antibody specifically detects PIKFyve/PIP5K3 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| PIKFyve/PIP5K3 | |
| Monoclonal | |
| Unconjugated | |
| NP_689884 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Sandwich ELISA | |
| 1D11 | |
| In 1x PBS, pH 7.4 | |
| 1-phosphatidylinositol-3-phosphate 5-kinase, EC 2.7.1.150, FLJ37746, FYVE finger-containing phosphoinositide kinase, KIAA0981CFD, MGC40423, p235, Phosphatidylinositol-3-phosphate 5-kinase, Phosphatidylinositol-3-phosphate 5-kinase type III, phosphoinositide kinase, FYVE finger containing, PIKfyve, PIP5K, PIP5K3FAB1, PIPkin-III, type III, Type III PIP kinase | |
| PIP5K3 (NP_689884, 342 a.a. ∽ 451 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LQSTEFSETPSPDSDSVNSVEGHSEPSWFKDIKFDDSDTEQIAEEGDDNLANSASPSKRTSVSSFQSTVDSDSAASISLNVELDNVNFHIKKPSKYPHVPPHPADQKGRR | |
| 0.1 mg | |
| Lipid and Metabolism | |
| 200576 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction