Learn More
Invitrogen™ Human PIM1, His Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
Specifications
Specifications
| Accession Number | P11309 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris HCl with 0.05% Triton X-100, 0.5 mM EDTA, 2 mM DTT, 50% glycerol, 500 mM NaCl and no preservative; pH 7.5 |
| Gene ID (Entrez) | 5292 |
| Molecular Weight (g/mol) | 40.7 kDa |
| Name | Human PIM1, His Tag |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
| Gene Alias | non-specific serine/threonine protein kinase; Oncogene PIM1; PIM; Pim1; Pim-1; pim-1 kinase 44 kDa isoform; pim-1 oncogene; pim-1 oncogene (proviral integration site 1); Pim-1 proto-oncogene, serine/threonine kinase; proto-oncogene serine/threonine-protein kinase Pim-1; proviral integration site 1; serine/threonine-protein kinase pim-1; serine-threonine kinase PIM-1 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.