missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIM2 Antibody (OTI5D5), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73412PECY55
This item is not returnable.
View return policy
Description
PIM2 Monoclonal specifically detects PIM2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| PIM2 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| PIM2 | |
| Full length recombinant protein of human PIM2 (NP_006866) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Flow Cytometry | |
| OTI5D5 | |
| Flow Cytometry | |
| EC 2.7.11.1, pim-2 oncogene, pim-2h, proto-oncogene Pim-2 (serine threonine kinase), serine/threonine protein kinase pim-2, serine/threonine-protein kinase pim-2 | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase | |
| 11040.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction