missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIM2 Monoclonal Antibody (OTI10A12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501064
This item is not returnable.
View return policy
Description
PIM proteins (PIM-1, PIM-2 and PIM-3) are oncogene-encoded serine/threonine kinases. PIM-2 is highly homologous to Pim-1 with similar oncogenic functions. PIM-2 is most abundantly expressed in hematopoietic tissues, spleen, thymus, and peripheral blood leukocytes, as well as in testis, small intestine, and colon. PIM-2 overexpression promotes resistance to a host of apoptotic stimuli; its expression is negatively regulated by growth factor depletion. Increased levels of PIM-2 has also been observed in certain cancers.Specifications
| PIM2 | |
| Monoclonal | |
| 0.84 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9P1W9 | |
| PIM2 | |
| Full length human recombit protein of human PIM2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI10A12 | |
| Unconjugated | |
| PIM2 | |
| Pim-2 proto-oncogene, serine/threonine kinase | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11040 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction