missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIM2 Monoclonal Antibody (OTI10A12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501064S
This item is not returnable.
View return policy
Description
PIM2 Monoclonal antibody specifically detects PIM2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| PIM2 | |
| Monoclonal | |
| 0.84 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9P1W9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11040 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI10A12 | |
| Unconjugated | |
| PIM2 | |
| DXCch3; PIM2; Pim-2; pim-2 oncogene; Pim-2 proto-oncogene, serine/threonine kinase; pim-2h; proto-oncogene Pim-2 (serine threonine kinase); proviral integration site 2; serine/threonine protein kinase pim-2; serine/threonine-protein kinase pim-2 | |
| Full length human recombinant protein of human PIM2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction