Learn More
PIMT Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-44294C
Product Code. 18794502
  • € 633.00 / 0.10mL
Description
PIMT Polyclonal antibody specifically detects PIMT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, Immunohistochemistry
DyLight 650
IgG
Polyclonal
The epitope recognized by this antibody maps to a region between residue 803 to 853 of human PRIP-Interacting Protein with Methyltransferase Domain using the numbering given in entry NP_079107.5 (GeneID 96764).