Learn More
PIMT Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44294C
This item is not returnable.
View return policy
Description
PIMT Polyclonal antibody specifically detects PIMT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Western Blot, Immunohistochemistry | |
| DyLight 650 | |
| IgG |
| Polyclonal | |
| The epitope recognized by this antibody maps to a region between residue 803 to 853 of human PRIP-Interacting Protein with Methyltransferase Domain using the numbering given in entry NP_079107.5 (GeneID 96764). |