missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIMT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110481
This item is not returnable.
View return policy
Description
Immunogen sequence: RVIAIDIDPV KIALARNNAE VYGIADKIEF ICGDFLLLAS FLKADVVFLS PPWGGPDYAT AETFDIRTMM SPDGFEIFRL SKKITNNIVY FLPRNADIDQ VASLAGPGGQ VEIEQNFLNN KLKTITAYFG DLIRRPASET.
Catalyzes the 2 serial methylation steps for the conversion of the 7-monomethylguanosine (m(7)G) caps of snRNAs and snoRNAs to a 2,2,7-trimethylguanosine (m(2,2,7)G) cap structure. The enzyme is specific for guanine, and N7 methylation must precede N2 methylation. Hypermethylation of the m7G cap of U snRNAs leads to their concentration in nuclear foci, their colocalization with coilin and the formation of canonical Cajal bodies (CBs). Plays a role in transcriptional regulation. [UniProt].
Specifications
| PIMT | |
| Polyclonal | |
| Unconjugated | |
| TGS1 | |
| Cap-specific guanine-N2 methyltransferase; CLL-associated antigen KW-2; D4Ertd800e; HCA137; Hepatocellular carcinoma-associated antigen 137; LOW QUALITY PROTEIN: trimethylguanosine synthase; NCOA6IP; nuclear receptor coactivator 6 interacting protein; nuclear receptor coactivator 6-interacting protein; Pimt; PIPMT; PRIP-interacting protein with methyltransferase motif; TGS1; tgs1 {ECO:0000250; Trimethylguanosine synthase; trimethylguanosine synthase 1; trimethylguanosine synthase homolog; trimethylguanosine synthase homolog (S. cerevisiae); trimethylguanosine synthase-like protein; UniProtKB:Q96RS0} | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 116940, 96764 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.72 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q923W1, Q96RS0 | |
| TGS1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 714-853 of human TGS1 (NP_079107.6). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction