Learn More
Elma™ Pin Sets
Click to view available options
Model:
0001R
0021R
0158
0509
0613/0614
0741
0767
0772
Unit Size:
Each
Description
- Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
- Ergonomic handles
- Autoclavable
- Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
- Square openings in the side walls for mounting silicone holders
Specifications
Specifications
| Quantity | 10 Pins/Set |
| Type | Pin Set |
| For Use With (Equipment) | Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY |
| Model | 0001R |