missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIN1 Monoclonal Antibody (OTI4H1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812247S
This item is not returnable.
View return policy
Description
PIN1 Monoclonal antibody specifically detects PIN1 in Human samples. It is validated for Western BlotSpecifications
| PIN1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P33681 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 941 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4H1 | |
| Unconjugated | |
| CD80 | |
| 0610025L01Rik; D9Bwg1161e; DOD; EC 5.2.1.8; NIMA-interacting; peptidylprolyl cis/trans isomerase, NIMA-interacting 1; peptidylprolyl cis/trans isomerase, NIMA-interacting 1 a; peptidylprolyl cis/trans isomerase, NIMA-interacting 1 S homeolog; Peptidyl-prolyl cis-trans isomerase NIMA-interacting 1; peptidyl-prolyl cis-trans isomerase Pin1; pin1; pin1.S; pin1a; pin1-a; pin1-b; PPIase Pin1; prolyl isomerase; prolyl isomerase Pin1; protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1; protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1; rotamase Pin1; ubl5; XELAEV_18021616mg | |
| Full length human recombinant protein of human CD80 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction