Learn More
PIN4 Antibody (14A7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26381-100ul
 View more versions of this product
Product Code. 30033248
  • € 533.00 / 100µL
Description
PIN4 Monoclonal antibody specifically detects PIN4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PIN4
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry
14A7
Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
4 (parvulin), MGC138486, Parvulin-17, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin)
A synthesized peptide derived from Human PIN4 [UniProt Q9Y237]
100 μL
metabolism
5303
Store at -20 to -70C. Avoid freeze-thaw cycles.