missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIN4 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97273FR
This item is not returnable.
View return policy
Description
PIN4 Polyclonal antibody specifically detects PIN4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PIN4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| 4 (parvulin), MGC138486, Parvulin-17, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin) | |
| Produced in rabbits immunized with E. coli-derived Human PIN4 fragment. | |
| 0.1 mL | |
| metabolism | |
| 5303 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction