missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIN4 Antibody (OTI7H2), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73416C
This item is not returnable.
View return policy
Description
PIN4 Monoclonal specifically detects PIN4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PIN4 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PIN4 | |
| Full length recombinant protein of human PIN4 (NP_006214) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI7H2 | |
| Western Blot, Immunohistochemistry | |
| 4 (parvulin), MGC138486, Parvulin-17, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin) | |
| Mouse | |
| Affinity Purified | |
| metabolism | |
| 5303.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction