missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PINCH1/LIMS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 624.00
Specifications
| Antigen | PINCH1/LIMS1 |
|---|---|
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18281392
|
Novus Biologicals
NBP2-56112 |
100 μL |
€ 624.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18628136
|
Novus Biologicals
NBP2-56112-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PINCH1/LIMS1 Polyclonal specifically detects PINCH1/LIMS1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PINCH1/LIMS1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| LIM and senescent cell antigen-like domains 1, LIM and senescent cell antigen-like-containing domain protein 1, PINCH-1, PINCHPINCH1Particularly interesting new Cys-His protein 1, Renal carcinoma antigen NY-REN-48 | |
| LIMS1 | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 3987 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MTALQLKELSHSGLYRRRRDRPDSLRVNGLPEEELSNMANAL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title