missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pinin Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-55315AF647
This item is not returnable.
View return policy
Description
Pinin Polyclonal antibody specifically detects Pinin in Human, Mouse samples. It is validated for Western Blot
Specifications
| Pinin | |
| Polyclonal | |
| Western Blot | |
| 140 kDa nuclear and cell adhesion-related phosphoprotein, Desmosome-associated protein, Domain-rich serine protein, DRS protein, DRSP, DRSSR-like protein, Melanoma metastasis clone A protein, MEMA, neutrophil protein, Nuclear protein SDK3, pinin, pinin, desmosome associated protein, SDK3 | |
| The immunogen recognized by this antibody maps to a region between residue 667 and the C-terminus (residue 717) of human Pinin, desmosome associated protein using the numbering given in entry NP_002678.2 (GeneID 5411). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 5411 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction