missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PINK1 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-493H
This item is not returnable.
View return policy
Description
PINK1 Polyclonal specifically detects PINK1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| PINK1 | |
| Polyclonal | |
| Western Blot | |
| Q9BXM7 | |
| PINK1 | |
| PINK1 antibody was developed using an N-terminal region synthetic peptide made to the human PINK1 protein sequence (between residues 1-50). [UniProt# Q9BXM7] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| PINK1 Antibody is expected recognize isoform 1 and basted on a homology blast, will not recognize isoform 2. | |
| Store at 4C in the dark. |
| Western Blot | |
| HRP | |
| PBS with No Preservative | |
| BRPK, EC 2.7.11.1, FLJ27236, PARK6, Parkinson disease (autosomal recessive) 6, protein kinase BRPK, PTEN Induced Kinase 1, PTEN induced putative kinase 1, PTEN-induced putative kinase protein 1, serine/threonine-protein kinase PINK1, mitochondrial | |
| Rabbit | |
| 63 kDa | |
| 0.1 mL | |
| Alzheimers Research, Cardiovascular Biology, DNA Repair, Lipid and Metabolism, Mitochondrial Markers, Mitophagy, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Kinase, Signal Transduction | |
| 65018 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction