missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PINK1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49678JF646
This item is not returnable.
View return policy
Description
PINK1 Polyclonal antibody specifically detects PINK1 in Human, Mouse samples. It is validated for Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| PINK1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| BRPK, EC 2.7.11.1, FLJ27236, PARK6, Parkinson disease (autosomal recessive) 6, protein kinase BRPK, PTEN Induced Kinase 1, PTEN induced putative kinase 1, PTEN-induced putative kinase protein 1, serine/threonine-protein kinase PINK1, mitochondrial | |
| PINK1 antibody was developed using a genomic peptide made to an internal region of the human PINK1 protein (within residues 350-500). [Swiss-Prot Q9BXM7] | |
| 0.1 mL | |
| Alzheimers Research, Autophagy, Cardiovascular Biology, DNA Repair, Lipid and Metabolism, Mitochondrial Markers, Mitophagy, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Kinase, Signal Transduction | |
| 65018 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction