missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PINX1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32265AF750
This item is not returnable.
View return policy
Description
PINX1 Polyclonal antibody specifically detects PINX1 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Western Blot, Immunohistochemistry | |
| The immunogen this antibody was made to, maps to a region between residue 225 to 275 of human PIN2 Interacting Protein 1 using the numbering given in entry NP_060354.4 (GeneID 54984). | |
| RUO | |
| Primary | |
| IgG |
| Rabbit | |
| Immunogen affinity purified | |
| Cancer | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction