missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PINX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32265
This item is not returnable.
View return policy
Description
PINX1 Polyclonal specifically detects PINX1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| PINX1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| FLJ20565, hepatocellular carcinoma-related putative tumor suppressor, Liver-related putative tumor suppressor, LPTLPinX1, LPTS67-11-3 protein, MGC8850, PIN2 interacting protein 1, PIN2/TERF1 interacting, telomerase inhibitor 1, PIN2/TERF1-interacting telomerase inhibitor 1, PIN2-interacting protein 1, Pin2-interacting protein X1, Protein 67-11-3, TRF1-interacting protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:1000 - 1:5000, Immunohistochemistry, Immunoprecipitation 2 - 10 ug/mg lysate | |
| NP_060354.4 | |
| PINX1 | |
| The immunogen this antibody was made to, maps to a region between residue 225 to 275 of human PIN2 Interacting Protein 1 using the numbering given in entry NP_060354.4 (GeneID 54984). | |
| 100 μL | |
| Cancer | |
| 54984 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction