Learn More
Invitrogen™ PINX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139773
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ENETTTTSAF TIQEYFAKRM AALKNKPQVP VPGSDISETQ VERKRGKKRN KEATGKDVES YLQP Highest antigen sequence indentity to the following orthologs: Rat - 63%, Mouse - 70%.
Microtubule-binding protein essential for faithful chromosome segregation. Mediates TRF1 and TERT accumulation in nucleolus and enhances TRF1 binding to telomeres. Inhibits telomerase activity. May inhibit cell proliferation and act as tumor suppressor. [UniProt].
Specifications
| PINX1 | |
| Polyclonal | |
| Unconjugated | |
| PINX1 | |
| 2210403I16Rik; 2610028A01Rik; 67-11-3; 67-11-3 protein; AU024023; EMBL:ABO28828.1}; FLJ20565; hepatocellular carcinoma-related putative tumor suppressor; liver-related putative tumor suppressor; LPTL; Lpts; LPTS1; MGC8850; PIN2 interacting protein 1; PIN2/TERF1 interacting, telomerase inhibitor 1; PIN2/TERF1-interacting telomerase inhibitor 1; PIN2/TRF1-interacting protein; pin2-interact; PIN2-interacting protein 1; PIN2-interacting protein X1; Pinx1; pinx1 {ECO:0000312; Protein 67-11-3; RGD1566025; TRF1-interacting protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54984 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96BK5 | |
| PINX1 | |
| Recombinant protein corresponding to Human PINX1. Recombinant protein control fragment (Product #RP-108747). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.