missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIP Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98683AF350
This item is not returnable.
View return policy
Description
PIP Polyclonal antibody specifically detects PIP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PIP | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 350 | |
| GCDFP15prolactin-inducible protein, GCDFP-15SABP, GPIP4gp17, Gross cystic disease fluid protein 15, prolactin-induced proteinSecretory actin-binding protein | |
| Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human PIP. | |
| 0.1 mL | |
| Cancer, Cellular Markers, Signal Transduction | |
| 5304 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction