missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIP Antibody (SR1245), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22064-50ul
This item is not returnable.
View return policy
Description
PIP Monoclonal antibody specifically detects PIP in Human samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation
Specifications
| PIP | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| SR1245 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| GCDFP15prolactin-inducible protein, GCDFP-15SABP, GPIP4gp17, Gross cystic disease fluid protein 15, prolactin-induced proteinSecretory actin-binding protein | |
| A synthesized peptide derived from human PIP (Uniprot #: P12273) | |
| 50 μg | |
| Cancer, Cellular Markers, Signal Transduction | |
| 5304 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction