Learn More
PIP4K2C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17077-1-AP-150UL
 View more versions of this product
Product Code. 16806914
  • € 410.00 / 150µL
Description
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.
Specifications
Specifications
PIP4K2C
Polyclonal
Unconjugated
PIP4K2C
PI(5)P 4 kinase type II gamma, PIP4K2C, PIP4KII gamma, PIP5K2C
Rabbit
Antigen Affinity Chromatography
RUO
117150, 140607, 79837
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O88370, Q8TBX8, Q91XU3
PIP4K2C
PIP4K2C Fusion Protein Ag10989
150 μL
Primary
Human, Mouse, Rat