Learn More
Brady™ Pipe Markers
Product Code. 17343410 Shop All Brady Products
Change view
Click to view available options
Display Text:
Argon
Hydrogen
Nitrogen
Oxygen
Unit Size:
Pack of 30
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Unit Size
17343410 Oxygen Pack of 30
17313410 Argon Pack of 30
17323410 Nitrogen Pack of 30
17333410 Hydrogen Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17343410 Supplier Brady™ Supplier No. N004529
  • ONLINE EXCLUSIVE PRICE
    € 128.00
    € 114.51 / Pack of 30
    Save € 13.49
    10% Off
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Individual pipe markers on a roll with perforated arrows and symbols.
  • Pipe and valve marking
  • Permanent acrylic based adhesive
Specifications
Adhesive Type Permanent Acrylic
Color Black on Yellow
Material Polyester
Display Text Oxygen
Dimensions (L x W) 355 x 37 mm
Alternate Part Number ZUURSTOF355X37RL-T1-LL-P2