missing translation for 'onlineSavingsMsg'
Learn More

Pipequaline, MedChemExpress

Product Code. 16419486
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16419486 50 mg 50mg
16484236 100 mg 100mg
16455537 5 mg 5mg
16422798 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16419486 Supplier Medchem Express Supplier No. HY100140

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pipequaline (PK 8165) is a partial benzodiazepine receptor agonist with anxiolytic activity.

Pipequaline (PK 8165) is a partial benzodiazepine receptor agonist with anxiolytic activity.

Specifications

Chemical Name or Material Pipequaline
CAS 77472-98-1
Color White
Purity Grade Notes Research
Molecular Formula C22H24N2
Quantity 50 mg
Synonym PK-8165
Solubility Information DMSO : ≥ 32 mg/mL (101.13 mM)
SMILES C1(C2=CC=CC=C2)=NC3=CC=CC=C3C(CCC4CCNCC4)=C1
Molecular Weight (g/mol) 316.44
Formula Weight 316.44
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.