missing translation for 'onlineSavingsMsg'
Learn More

Piperonyloyl chloride, 98%

Product Code. p-2692064 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10633003 1 g Glass bottle 1g
10314362 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10633003 Supplier Thermo Scientific Acros Supplier No. 370790010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25054-53-9 | C8H5ClO3 | 184.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25054-53-9
Molecular Formula C8H5ClO3
Molecular Weight (g/mol) 184.58
MDL Number MFCD00016904
InChI Key ZRSGZIMDIHBXIN-UHFFFAOYSA-N
Synonym piperonyloyl chloride, benzo d 1,3 dioxole-5-carbonyl chloride, 2h-1,3-benzodioxole-5-carbonyl chloride, piperonoyl chloride, benzo 1,3 dioxole-5-carbonyl chloride, acmc-20ajvo, piperonylic acid chloride, 3,4-methylenedioxybenzoylchloride, 3,4-methylenedioxybenzoyl chloride
PubChem CID 2734749
IUPAC Name 1,3-benzodioxole-5-carbonyl chloride
SMILES C1OC2=C(O1)C=C(C=C2)C(=O)Cl

Specifications

Melting Point 77°C to 81°C
Color Blue-Gray to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 184.58
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Piperonyloyl chloride
compliance-icons
Product Identifier
  • Piperonyloyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.