Learn More
SymLine™ Straight Pipe Systems
Product Code. 14776901
Change view
Click to view available options
Length (Metric):
300 mm
900 mm
1200 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
14776901 300 mm Each
14786901 900 mm Each
14796901 1200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 14776901 Supplier SymLine™ Supplier No. 106435
The SymLine™ pipe system provides connection to the central collection point. Place the collecting container or canister where you want it: the pipe system can be exactly configured to suit your needs!
  • Straight pipes are available in multiple standard lengths
  • A wide range of connectors, branches and manifolds allows the pipe system to be connected precisely to the conditions in your laboratory
Specifications
Diameter (Metric) Outer 32 mm
Length (Metric) 300 mm
For Use With (Application) Connecting Pipes with a 32 mm OD
For Use With (Equipment) Tube System with SymLine, FlexTube and FlexTubePro, Connectors, Distributors and Shut-off
Material HDPE-EC
Product Type Connection for Pipe System