Learn More
Greiner Bio-One 10 mL Sterile Serological Pipets
Drip-free pipetting design with high optical clarity and maximum accuracy.
Brand: Greiner Bio-One 607107
Additional Details : Weight : 3.69000kg
Description
- High-grade polystyrene
- Clear, easily legible graduation
- Colored vertical stripe for ease in reading the volume
- With synthetic fibrous filter material for protection against
Specifications
Shape: Standard | |
Sterile | |
1/10 | |
10 mL |
Polystyrene | |
Yes | |
Bulk | |
Serological Pipet |