Learn More
Reitenspiess™ Pipet Brush
Product Code. 10549431
Product Code. 10549431
Brand: Reitenspiess™ 10
Only null left
Add to basket
Edge
Add to basket
Use the two different brush widths of the Reitenspiess™ Pipet Brush to allow the simultaneous cleaning of delivery tubes and body. A flexible twisted wire handle with hanging loop secures the bristles.
  • Two different brush widths
  • Simultaneous cleaning of delivery tubes and body 
  • Flexible twisted wire handle with hanging loop
Specifications
Type Pipette Brush