Learn More
Mettler Toledo™ Pipet-Lite™ XLS Model 8-Channel Variable Volume Manual Pipette
Product Code. 11372325
Encompass
Change view
Click to view available options
Volume (Metric):
1200 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11372325 1200 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11372325 Supplier Mettler Toledo™ Supplier No. 17011842
This item requires a price update prior to being ordered.
Request a Price for this item here.
Specifications
Model XLS+
Number of Channels 8
Product Line Pipet-Lite™ XLS
Type Variable Volume Manual Pipette
Volume (Metric) 1200 μL
For Use With (Application) Genomic, Proteomic, Tissue Culture and Cell Culture applications
Quantity 1 Pack