Learn More
Sartorius Pipet Stands and Accessories
Optimize the use of mLINE™ and Proline™ mechanical pipets. Sartorius Pipet Stands and Accessories include single- and multi-pipet stands, colored-coded caps and a calibration tool. Can also be used with a range of standard pipet brands.
Brand: Sartorius 726001
Additional Details : Weight : 0.00200kg
Description
Pipette Stands- Stores pipettes in an upright position while not in use to avoid contamination from the desktop
- Linear Stand is designed for all mechanical devices and stores them neatly in a line for instant access
- Carousel stand is ideal for saving bench space and stores up to 6 mechanical pipettes
- mLINE/Proline Plus Single Holder compactly holds a single mechanical pipette
- Colored caps help identify the volume of mLINE pipettes so it can easily be matched to the corresponding tip size
- Includes 5 color-coded replacement caps
- Calibration Tool for mLINE or Proline Plus mechanical pipettes for continuous accuracy and precision
- Can also be used as a tube opener
Specifications
Color Coded Caps | |
Color coding mLINE pipets by volume | |
5/Pk. |
Gray/Yellow/Blue/Green/Red | |
mLINE™/Proline™ Plus mechanical pipette |