Learn More
AHN biotechnologie pipet4u™ pro Single-channel Variable-volume Pipette
Description
- Comfortable and effortless pipetting / user-friendly handle design with soft grip for comfortable operation to prevent hand stress / light weight / low plunger force minimizes RSI (repetitive strain injury).
- Easy identification - attachable color coding.
- Durability - tip cone demonstrates high mechanical strength and high chemical resistance.
- Calibration stability - even after multiple autoclaving, no recalibration is required.
- Accurate and reliable - reverse pipetting option for accurate and precise aspirating and dispensing of viscous liquids / innovative soft grip prevents transfer of heat to internal components, ensuring accurate pipette operation / lock counter mechanism of variable volume pipettes prevents volume changes while pipetting / blow out system allows last drop dispensing.
- Sterile handling - fully autoclavable pipette (by steam at 121 °C / 15 min).
- Easy and quick maintenance and adjustment - with the included disassembly and calibration tool.
Specifications
Specifications
| Accuracy | 0.9-3.0% |
| Certifications/Compliance | DIN EN ISO 8655-1, DIN EN ISO 8655-2, DIN EN ISO 8655-6, EU Directive 98/79/E |
| Autoclavable | Autoclavable |
| Color | Assorted |
| Number of Channels | 1 |
| Product Line | Pipet4u |
| Sterility | Sterile |
| Type | Pipette |
| Volume (Metric) | 2 to 20 μL |
| Increments | 0.1 μL |
| Show More |