missing translation for 'onlineSavingsMsg'
Learn More

AHN biotechnologie pipet4u™ pro Single-channel Variable-volume Pipette

Product Code. 15868637
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
0.5 to 5 mL
100 to 1000 μL
2 to 20 μL
2.0 to 10 mL
20 to 200 μL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15868637 0.5 to 5 mL Each
15848637 0.5 to 10 μL Each
15878637 2.0 to 10 mL Each
15858637 2 to 20 μL Each
15832952 20 to 200 μL Each
15899512 100 to 1000 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15868637 Supplier AHN biotechnologie Supplier No. 8107209

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Comfortable, accurate, reliable, and autoclavable pipette.

  • Comfortable and effortless pipetting / user-friendly handle design with soft grip for comfortable operation to prevent hand stress / light weight / low plunger force minimizes RSI (repetitive strain injury).
  • Easy identification - attachable color coding.
  • Durability - tip cone demonstrates high mechanical strength and high chemical resistance.
  • Calibration stability - even after multiple autoclaving, no recalibration is required.
  • Accurate and reliable - reverse pipetting option for accurate and precise aspirating and dispensing of viscous liquids / innovative soft grip prevents transfer of heat to internal components, ensuring accurate pipette operation / lock counter mechanism of variable volume pipettes prevents volume changes while pipetting / blow out system allows last drop dispensing.
  • Sterile handling - fully autoclavable pipette (by steam at 121 °C / 15 min).
  • Easy and quick maintenance and adjustment - with the included disassembly and calibration tool.

Specifications

Accuracy 0.5-2.0%
Certifications/Compliance DIN EN ISO 8655-1, DIN EN ISO 8655-2, DIN EN ISO 8655-6, EU Directive 98/79/E
Autoclavable Autoclavable
Color Assorted
Number of Channels 1
Product Line Pipet4u
Sterility Sterile
Type Pipette
Volume (Metric) 0.5 to 5 mL
Increments 0.05 mL
Precision 0.15-0.60%
Warranty 2 Years
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.