Learn More
VITLAB™ Pipette Fillers
Measure the volume of liquids using Vitlab™ Pipette Filters. These filters draw liquid into the pipette.
€ 8.50 - € 173.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10489382
|
VITLAB™
104099 |
Up to 10mL Volumetric or Measuring Pipettes |
€ 8.50
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
10333681
|
VITLAB™
002516 |
2mL Pipettes, Glass and Plastic Pipettes |
€ 17.00
Each |
In Stock
Log in to see stock. |
|||||
16513642
|
VITLAB™
324594 |
2mL Pipettes, Glass and Plastic Pipettes |
€ 173.00
Pack of 10 |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
10644222
|
VITLAB™
324594 |
2mL Pipettes, Glass and Plastic Pipettes |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Slow rotation of the actuator-wheels draws liquid into the pipette
- Pressing air bleed valve automatically empties the pipette without returning the piston
- Green, Blue and Red available