missing translation for 'onlineSavingsMsg'
Learn More

Terracycle™ Pipette Tip Boxes Zero Waste Box™

Product Code. 17845805
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17845805 Small Each
17805805 Medium Each
17825795 Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17845805 Supplier Terracycle™ Supplier No. CS180SNL
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recycle Pipette Tip Boxes using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.

Use this Zero Waste Box to recycle:

  • Empty pipette tip boxes

Do not send in:

  • Pipette tips, tubes and rigid lab plastics that are contaminated with materials considered hazardous or dangerous
  • Autoclaved material
  • Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
  • Batteries, pressurized canisters, broken glass and medical waste

Important:

  • We do not collect any hazardous waste containing substances with toxic, reactive, corrosive, infectious, flammable, explosive properties or harmful to human health and the environment.
  • Please make sure that you do not mix hazardous waste with non-hazardous waste.

Specifications

Capacity (Metric) 7 kg
Capacity (English) 15 lb.
Color Beige, Green, Red, White
Dimensions (L x W x H) 25 x 25 x 46 cm
Disposable Single-use
Latex Free Yes
Size Small
Sterility Non-sterile
Autoclavable Not Autoclavable
Material Cardboard, Plastic
Product Type Recycling Box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.