missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Pipette Tip, Racked, Sterile

Product Code. 32035889 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
300 μL
1000 μL
1200 μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric)
32035889 10 μL
32035930 200 μL
32035964 300 μL
32035907 1000 μL
32035873 1200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 32035889 Supplier Axygen™ Supplier No. PT300RS
  • ONLINE EXCLUSIVE PRICE
    € 63.15
    € 43.97 / Pack of 960
    Save € 19.18
    30% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

RNase-/DNase-free

  • For more than 20 years Axygen Universal pipette tips have set the standard for quality accuracy and precision in laboratories worldwide
  • Redesigned to meet every researcher needs the next generation of Axygen Universal Fit pipette tips deliver improved pipettor compatibility durable hinged racks and a complete offering from 10 μL to 1200 μL
  • Features include extended length tips wide bore designs for viscous solutions and proprietary MaxRecovery+™ technology to enhance liquid handling performance
  • RNaseDNase free
  • Nonpyrogenic
  • Human DNA free
  • ATP free
  • Available with MaxRecovery+ proprietary next generation low retention technology formulated with no intentionally added PFAS
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Format Rack
Tip Style Fine Point
Quantity 4800/Cs., 960/Pk.
Product Type Pipette Tips
Color Code Yellow
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene
Fit for Purpose Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.