Learn More
VistaLab™ Technologies Pipetter Nozzles
Contains an internal aerosol/liquid barrier filter to prevent liquid from being aspirated into pipetter
Brand:  VistaLab™ Technologies 9060-1010
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11986032
  • € 82.72 / Pack of 4
Estimated Shipment: 01-07-2024
to see stock.
Add to basket
Description
Description
  • Available in range of sizes
Specifications
Specifications
Non-sterile
0.5 to 20 μL Ovation™ BioNatural™ Multichannel Pipette
2 in.
5 in.
0.5 to 20 μL
4/Box
2 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers