missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ 1000μL Conductive Non-Sterile CO-RE Pipet Tips with Filter

Product Code. 13477697
Change view
Click to view available options
Volume (Metric):
1000 μL
Unit Size:
Pack of 3840
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
13477697 1000 μL Pack of 3840
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 13477697 Supplier Hamilton™ Supplier No. 235905_CORE_I
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Essential for precise pipetting. Hamilton™ 1000μL Conductive Non-Sterile Filter Pipet Tips provide enhanced pipetting safety and reliability with the CO-RE PerfectTouch system.

  • 95.1mm length
  • Color code label: white
  • Usable with 1000μL pipetting channels/CO-RE 96 Probe Head
  • Pressure-based Liquid Level Detection (pLLD) and capacitive (cLLD) with conductive (black) version
  • Process safety offered by Monitored Air Displacement and Total Aspiration and Dispense Monitoring

Specifications

Volume (Metric) 1000 μL
Filtered Filtered
Sterility Non-sterile
Color Black
Rack Color White
Color Code White
For Use With (Equipment) 1000 μL Pipetting Channels/CO-RE 96 Probe Head
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.