Learn More
Hamilton™ 300μL Conductive CO-RE Pipet Tips in Nested Racks
Essential for precise pipetting. Hamilton™ 300μL Conductive CO-RE Pipet Tips in Nested Racks provide enhanced pipetting safety and reliability with the CO-RE PerfectTouch system.
Brand:  Hamilton™ 235950
Additional Details : Weight : 9.99000kg
Product Code. 12984320
  • € 1630.00 / Pack of 11520
In Stock
to see stock.
Add to basket
Description
Description
  • For use with the CO-RE 384 Probe Head
  • Fine, thin tip facilitates dosing of micro volumes
  • Capacitive LLD (cLLD) with conductive (black) version
  • Process safety offered by Monitored Air Displacement
Specifications
Specifications
300 μL
Non-sterile
Single-use
Non-Filtered
Black
CO-RE™ 384 Probe Head