Learn More
Hamilton™ 300μL Conductive CO-RE Pipet Tips in Nested Racks
Essential for precise pipetting. Hamilton™ 300μL Conductive CO-RE Pipet Tips in Nested Racks provide enhanced pipetting safety and reliability with the CO-RE PerfectTouch system.
Brand: Hamilton™ 235950
Additional Details : Weight : 9.99000kg
Description
- For use with the CO-RE 384 Probe Head
- Fine, thin tip facilitates dosing of micro volumes
- Capacitive LLD (cLLD) with conductive (black) version
- Process safety offered by Monitored Air Displacement
Specifications
300 μL | |
Non-sterile | |
Single-use |
Non-Filtered | |
Black | |
CO-RE™ 384 Probe Head |