Learn More
Kartell SpA Sterile Filter Tips in Rack
Product Code. 10010822
Change view
Click to view available options
Volume (Metric):
0.1 to 1 mL
0.5 to 10 μL
1 to 200 μL
1 to 30 μL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
Unit Size:
Pack of 960
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10010822 1 to 200 μL Pack of 960
10557051 100 to 1000 μL Pack of 960
10716352 10 to 100 μL Pack of 960
10276571 0.1 to 1 mL Pack of 960
10144571 1 to 30 μL Pack of 960
10104671 0.5 to 10 μL Pack of 960
10796722 2 to 20 μL Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10010822 Supplier Kartell SpA Supplier No. 0962900
Wastewater Testing Savings
Use promo code 28023
View all promotions
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
Sterile disposable pipette tips with filter, polypropylene
Made of polypropylene and including a filter, these tips are DNase, RNase and DNA free, ATP and metal free, cadmiuim free, apyrogenic and PCR inhibitor free. Sterilized by gamma radiation, they are autoclavable at +121°C for 20 minutes. Designed for use in molecular biology, cell cultures, bacteriology, virology, immunology and radioimmunology. Hydrophobic polyethylene filters act as a barrier against aerosol contamination, preventing cross-contamination of samples and micropipettes. Tip orifices are sized to completely empty the tip in the event of over-aspiration. Although recommended for use in automatic Kartell micropipettes, these tips deliver optimum results when used with most micropipette brands available on the market.
The indicated tip/micropipette compatibility may be subject to change, in case of any structural change carried out on micropipette models by manufacturers
Specifications
Volume (Metric) 1 to 200 μL
Filtered Filtered
Sterility Sterile
Color Transparent
Autoclavable Autoclavable
Format Rack, 10 x 96
Tip Style Low Retention
Product Type Filter Tips
Color Code White
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Virgin Cadmium-free Polypropylene
Fit for Purpose Sterile