missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIR-A/B Antibody (404127), DyLight 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB5308K
This item is not returnable.
View return policy
Description
PIR-A/B Monoclonal antibody specifically detects PIR-A/B in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| PIR-A/B | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Mouse myeloma cell line NS0-derived recombinant mouse PIR-A1, Gly24-Met650, Accession # A2NTJ8 | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 404127 | |
| Flow Cytometry, CyTOF-ready | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction