missing translation for 'onlineSavingsMsg'
Learn More

Piromidic acid, MedChemExpress

Product Code. 16418658
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16418658 10 mg 10mg
16474527 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16418658 Supplier Medchem Express Supplier No. HYB1043

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Piromidic acid is an antibacterial agent. Piromidic acid is active against gramnegative bacteria and staphylococci and can be used for the research of intestinal, urinary, and biliary tract infections.

Piromidic acid is an antibacterial agent. Piromidic acid is active against gramnegative bacteria and staphylococci and can be used for the research of intestinal, urinary, and biliary tract infections.

Specifications

Chemical Name or Material Piromidic acid
Physical Form Solid
CAS 19562-30-2
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C14H16N4O3
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 1 mg/mL (3.47 mM; Need ultrasonic) ∣H2O : 1 mg/mL (3.47 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(C1=CN(CC)C2=NC(N3CCCC3)=NC=C2C1=O)O
Molecular Weight (g/mol) 288.3
Formula Weight 288.3
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.