missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PISD Antibody (OTI4G5), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73420AF700
This item is not returnable.
View return policy
Description
PISD Monoclonal specifically detects PISD in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PISD | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PISD | |
| Recombinant protein fragment corresponding to amino acids 80-340 of human PISD (NP_055153) produced in E.coli. | |
| 0.1 mL | |
| 23761.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI4G5 | |
| Western Blot, Immunohistochemistry | |
| DJ858B16, dJ858B16.2, DKFZp566G2246, EC 4.1.1.65, phosphatidylserine decarboxylase, phosphatidylserine decarboxylase proenzyme, PSD, PSDC, PSSC | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction