missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PISD Monoclonal Antibody (OTI2H1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807331
This item is not returnable.
View return policy
Description
Phosphatidylserine decarboxylases catalyze the formation of phosphatidylethanolamine (PE) by decarboxylation of phosphatidylserine (PS). Type I PSDs, such as PISD, are targeted to the inner mitochondrial membrane by an N-terminal targeting sequence. PISD also contains a conserved LGST motif that functions as an autocatalytic cleavage site where the proenzyme is split into mature alpha and beta subunits.Specifications
| PISD | |
| Monoclonal | |
| Unconjugated | |
| PISD | |
| DJ858B16, PSD, PSDC, PSSC, dJ858B16.2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23761 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI2H1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UG56 | |
| PISD | |
| Human recombit protein fragment corresponding to amino acids 80-340 of human PISD produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction