missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PISD Monoclonal Antibody (OTI2H1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807331S
This item is not returnable.
View return policy
Description
PISD Monoclonal antibody specifically detects PISD in Human samples. It is validated for Western BlotSpecifications
| PISD | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UG56 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23761 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI2H1 | |
| Unconjugated | |
| PISD | |
| 9030221M09Rik; DJ858B16; dJ858B16.2; DKFZp566G2246; phosphatidylserine decarboxylase; Phosphatidylserine decarboxylase alpha chain; Phosphatidylserine decarboxylase beta chain; phosphatidylserine decarboxylase proenzyme; phosphatidylserine decarboxylase proenzyme, mitochondrial; phosphatidylserine decarboxylase proenzyme; phosphatidylserine decarboxylase proenzyme, mitochondrial; Pisd; PSD; PSDC; PSSC | |
| Human recombinant protein fragment corresponding to amino acids 80-340 of human PISD produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction